Learn More
BTNX Inc BTNX Urinalysis Reagent Test Strip 5-Parameter (Glucose Protein Nitrite Blood Leukocytes) 100 Strips/Bottle
Catalog No. NC3957496
Encompass_Preferred
The Rapid Response 5-Parameter Urinalysis Reagent Strips are qualitative and semi-qualitative dip-and-read test for the detection of urine analytes in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases.The 5 Parameter Rapid Response Urinalysis Reagent Strips simultaneously detects Glucose, Protein, Nitrite, Blood and Leukocytes.
Catalog No. NC3957496 Supplier BTNX Inc Supplier No. U5.11S100
The Rapid Response 5-Parameter Urinalysis Reagent Strips are qualitative and semi-qualitative dip-and-read test for the detection of urine analytes in human urine, to aid in the general evaluation of health and in the diagnosis and monitoring of metabolic or systemic diseases.The 5 Parameter Rapid Response Urinalysis Reagent Strips simultaneously detects Glucose, Protein, Nitrite, Blood and Leukocytes.