Learn More
BTNX Inc BTNX Urinary Tract Infection Test Strip 2-Parameter (LEU/NIT) 3 Tests/Kit
Catalog No. NC3957502
Encompass_Preferred
The Rapid Response Urinary Tract Infection Test Strips tests for leukocytes and nitrite in urine to aid in the screening of urinary tract infection (UTI). This test can be given to patients to check for an acute urinary tract infection, and to help monitor and assess therapy treatment. Quantity: 3/kit. Time to result: 1-2 minutes.
Catalog No. NC3957502 Supplier BTNX Inc Supplier No. UTI1S1
The Rapid Response Urinary Tract Infection Test Strips tests for leukocytes and nitrite in urine to aid in the screening of urinary tract infection (UTI). This test can be given to patients to check for an acute urinary tract infection, and to help monitor and assess therapy treatment. Quantity: 3/kit. Time to result: 1-2 minutes.