Learn More
ACROBiosystems Biotinylated Cynomolgus Transferrin Receptor protein, His-tagged, 200 μg, >95% purity, Dimer, 78.8 kDa
Catalog No. NC3356521
Encompass_Preferred
Biotinylated Cynomolgus Transferrin Receptor protein, His-tagged, 200 μg, >95% purity, Dimer, 78.8 kDa
Catalog No. NC3356521 Supplier ACROBiosystems Supplier No. TFRC82E3200UG
Biotinylated Cynomolgus Transferrin Receptor protein, His-tagged, 200 μg, >95% purity, Dimer, 78.8 kDa