Learn More
Cole-Parmer Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC
Catalog No. NC1053599 Shop All Cole-Parmer Products
Encompass_Preferred
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100C with ±1C digital control.
Catalog No. NC1053599 Supplier Cole-Parmer Supplier No. 4417500
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100C with ±1C digital control.