Learn More
Medchemexpress LLC HY-111617 10mg Medchemexpress, BTR-1 CAS:18331-34-5 Purity:>98%
SDP
Catalog No. 501968927
Encompass_Preferred
Medchemexpress, HY-111617 10mg BTR-1 CAS:18331-34-5 BTR-1 is an active anti-cancer agent, causes S phase arrest, and affects DNA replication in leukemic cells. BTR-1 activates apoptosis and induces cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8927 Supplier Medchemexpress LLC Supplier No. HY11161710MG
Medchemexpress, HY-111617 10mg BTR-1 CAS:18331-34-5 BTR-1 is an active anti-cancer agent, causes S phase arrest, and affects DNA replication in leukemic cells. BTR-1 activates apoptosis and induces cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.