Learn More
Medchemexpress LLC BTR2000 TFA | 99.0% | 764.83 | 5 MG
SDP
Catalog No. 5000364123
Encompass_Preferred
BTR2000 TFA is a specifically KLHL20 ligand that binds to the KLHL20 Kelch domain with high affinity. It can be utilized as a ligand for E3 ligase in the synthesis of BTR2004.
  • Specifically targets KLHL20
  • Binds to the KLHL20 Kelch domain with high affinity
  • Useful as a ligand for E3 ligase
  • Used for the synthesis of BTR2004
Catalog No. 50-003-64123 Supplier Medchemexpress LLC Supplier No. 5MGHY172563A
BTR2000 TFA is a specifically KLHL20 ligand that binds to the KLHL20 Kelch domain with high affinity. It can be utilized as a ligand for E3 ligase in the synthesis of BTR2004.
  • Specifically targets KLHL20
  • Binds to the KLHL20 Kelch domain with high affinity
  • Useful as a ligand for E3 ligase
  • Used for the synthesis of BTR2004