Learn More
Medchemexpress LLC HY-124754 50mg , BTRX-335140 CAS:2244614-14-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12475450MG
Encompass_Preferred
HY-124754 50mg BTRX-335140 CAS: 2244614-14-8 BTRX-335140 (CYM-53093) is a selective and orally active κ opioid receptor (KOR) antagonist, has antagonist activity for κOR, μOR and δOR with IC50 values of 0.8 nM, 110 nM, and 6500 nM, respectively.BTRX-335140 endows with favorable in vitro ADMET and in vivo pharmacokinetic profiles and medication-like duration of action in rats. BTRX-335140 distributes well into the CNS and can be used for the research of neuropathy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0634