Learn More
Apexbio Technology LLC BTS 1576-37-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000090763
Encompass_Preferred
BTS (1576-37-0) is a small-molecule inhibitor targeting skeletal muscle myosin II subfragment 1 (S1) ATPase It is designed to inhibit S1 ATPase activity thereby modulating skeletal muscle contraction by selectively suppressing actin- and Ca2 -stimulated ATP hydrolysis in fast muscle fibers BTS exerts its biological activity primarily through ATPase inhibition In in vitro studies BTS demonstrates selective inhibitory activity with an IC50 value of approximately 5 M against skeletal muscle myosin S1 ATPase Based on these pharmacological properties BTS holds research potential in delineating skeletal muscle contraction-relaxation mechanisms characterizing sarcomere movement and studying metabolic pathways underlying skeletal muscle activity as well as in evaluating pharmacological interventions affecting muscle function
Catalog No. 50-000-90763 Supplier Apexbio Technology LLC Supplier No. B47125.1
BTS (1576-37-0) is a small-molecule inhibitor targeting skeletal muscle myosin II subfragment 1 (S1) ATPase It is designed to inhibit S1 ATPase activity thereby modulating skeletal muscle contraction by selectively suppressing actin- and Ca2 -stimulated ATP hydrolysis in fast muscle fibers BTS exerts its biological activity primarily through ATPase inhibition In in vitro studies BTS demonstrates selective inhibitory activity with an IC50 value of approximately 5 M against skeletal muscle myosin S1 ATPase Based on these pharmacological properties BTS holds research potential in delineating skeletal muscle contraction-relaxation mechanisms characterizing sarcomere movement and studying metabolic pathways underlying skeletal muscle activity as well as in evaluating pharmacological interventions affecting muscle function