Learn More
Cayman Chemical BTS-54-505 (hydrochloride), Purity: ≥98%, CAS: 84484-78-6, 10 mg
SDP
Catalog No. 502941977
Encompass_Preferred
An active metabolite of sibutramine; inhibits norepinephrine, 5-HT, and dopamine uptake in rat cortical synaptosomes (IC50s = 0.066, 5.1, and 0.61 µM, respectively); prevents reserpine-induced ptosis in rats (ED50 = 3.7 mg/kg); decreases immobility in the Porsolt learned helplessness test in mice at 10 mg/kg; increases colonic temperature and decreases food intake in rats at 10 mg/kg
Catalog No. 50-294-1977 Supplier Cayman Chemical Supplier No. 3982810 MG
An active metabolite of sibutramine; inhibits norepinephrine, 5-HT, and dopamine uptake in rat cortical synaptosomes (IC50s = 0.066, 5.1, and 0.61 µM, respectively); prevents reserpine-induced ptosis in rats (ED50 = 3.7 mg/kg); decreases immobility in the Porsolt learned helplessness test in mice at 10 mg/kg; increases colonic temperature and decreases food intake in rats at 10 mg/kg