Learn More
Selleck Chemical LLC BTSA1 2mg 314761-14-3
SDP
Supplier:  Selleck Chemical LLC S86502MG
Encompass_Preferred
BTSA1 is a pharmacologically optimized BAX activator that binds with high affinity and specificity to the N-terminal activation site and induces conformational changes to BAX leading to BAX-mediated apoptosis. It effectively promotes apoptosis in leukemia cell lines and patient samples while sparing healthy cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8157