Learn More
New England Biolabs, Inc. BtsIMutI – 100 units
SDP
Catalog No. 504466
Encompass
A restriction endonuclease that recognizes the sequence CAGTG_NN^.
Catalog No. 50-446-6 Supplier New England Biolabs, Inc. Supplier No. R0664S
A restriction endonuclease that recognizes the sequence CAGTG_NN^.