Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BTT-266 10mg | 2413939-89-4 | 10 MG
SDP

Catalog No. 5000227920
Encompass_Preferred

BTT-266 is a small-molecule calcium channel antagonist that suppresses the CaVα1-AID-CaVβ3 interaction (reported Ki = 1.4 μM). It is provided as a high-purity powder for biochemical and pharmacological research on voltage-dependent calcium channels.

  • High purity (98.90%) powder for research use.
  • Molecular weight 556.61 and formula C34H28N4O4.
  • Soluble in DMSO at 12.5 mg/mL; sonication may be required.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Suitable for biochemical assays and calcium channel modulation studies.

Catalog No. 50-002-27920 Supplier Medchemexpress LLC Supplier No. HY11559710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BTT-266 is a small-molecule calcium channel antagonist that suppresses the CaVα1-AID-CaVβ3 interaction (reported Ki = 1.4 μM). It is provided as a high-purity powder for biochemical and pharmacological research on voltage-dependent calcium channels.

  • High purity (98.90%) powder for research use.
  • Molecular weight 556.61 and formula C34H28N4O4.
  • Soluble in DMSO at 12.5 mg/mL; sonication may be required.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Suitable for biochemical assays and calcium channel modulation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.