Learn More
Medchemexpress LLC HY-100486 5mg Medchemexpress, BTTAA CAS:1334179-85-9 Purity:>98%
SDP
Catalog No. 501931850
Encompass_Preferred
Medchemexpress, HY-100486 5mg BTTAA CAS:1334179-85-9 BTTAA is a Cu(I)-stabilizing ligand, whch performs potently with ubiquitin Glu18AzF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1850 Supplier Medchemexpress LLC Supplier No. HY1004865MG
Medchemexpress, HY-100486 5mg BTTAA CAS:1334179-85-9 BTTAA is a Cu(I)-stabilizing ligand, whch performs potently with ubiquitin Glu18AzF. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.