Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bttaa

Catalog No. 906328500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-632-8500MG 100 mg
90-632-8100MG 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 90-632-8500MG Supplier Sigma Aldrich Supplier No. 906328500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

BTTAA is a next-generation, water-soluble ligand for the copper(I)-catalyzed azide-alkyne cycloaddition (CuAAC) that dramatically accelerates reaction rates and suppresses cell cytotoxicity. The biocompatibility and fast kinetics of BTTAA are advancements from water-insoluble TBTA and are desirable for bio conjugation in diverse chemical biology experiments.Biocompatible click chemistry enabled compartment-specific pH measurement inside E. coliFast, cell-compatible click chemistry with copper-chelating azides for biomolecular labelingMetabolic labeling of fucosylated glycoproteins in Bacteroidales speciesIncreasing the Efficacy of Bioorthogonal Click Reactions for Bioconjugation: A Comparative Study

Specifications

CAS 80279-50-1
Molecular Formula C19H30N10O2
Linear Formula C19H30N10O2
MDL Number MFCD29918287
Quantity 100 mg
Synonym 2-(4-((Bis((1-(tert-butyl)-1H-1,2,3-triazol-4-yl)methyl)amino)methyl)-1H-1,2,3-triazol-1-yl)acetic acid; Copper click-chemistry ligand; Water-soluble CuAAC ligand
Molecular Weight (g/mol) 430.51 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.