Learn More
Wilmad Labglass BTTLE GAS WASH FRTD COMP 250ML
Catalog No. 50805457
Encompass_Preferred
Bottle, Gas Washing, with Fritted Disc, Complete. Capacity: 250mL; Joint Size: [ST] 45/50. The lower Inlet tube has a coarse porosity disc sealed on to provide a stream of fine bubbles into the bottle.
Catalog No. 50-805-457 Supplier Wilmad Labglass Supplier No. LG3750102
Bottle, Gas Washing, with Fritted Disc, Complete. Capacity: 250mL; Joint Size: [ST] 45/50. The lower Inlet tube has a coarse porosity disc sealed on to provide a stream of fine bubbles into the bottle.