Learn More
Wilmad Labglass BTTLE GAS WASH TALL COMP 250ML
Catalog No. 50807122
Encompass_Preferred
Bottle, Gas Washing, Tall Form, Drechsel, Complete. Capacity: 250mL; Joint Size: [ST] 29/42. Complete unit is supplied with stopper having gas Inlet and outlet tubes
Catalog No. 50-807-122 Supplier Wilmad Labglass Supplier No. LG3690100
Bottle, Gas Washing, Tall Form, Drechsel, Complete. Capacity: 250mL; Joint Size: [ST] 29/42. Complete unit is supplied with stopper having gas Inlet and outlet tubes