Learn More
Chemglass Life Sciences Bottle, Wide Mouth, Amber, Convenience Pack, 53-400 Thread Size, 500mL/16oz, PTFE Lined Green Cap
SDP
Catalog No. 50872739
Encompass_Preferred
Bottle, Wide Mouth, Amber, Convenience Pack, 53-400 Thread Size, 500mL/16oz, PTFE Lined Green Cap. Amber bottles for protection of light sensitive materials. Large wide mouth opening provides for easier access of contents. Available with a choice of cap styles, PTFE lined, Polyseal, or Vinyl lined Pulp Supplied with caps attached in convenience packs.
Catalog No. 50-872-739 Supplier Chemglass Life Sciences Supplier No. CG82108
Bottle, Wide Mouth, Amber, Convenience Pack, 53-400 Thread Size, 500mL/16oz, PTFE Lined Green Cap. Amber bottles for protection of light sensitive materials. Large wide mouth opening provides for easier access of contents. Available with a choice of cap styles, PTFE lined, Polyseal, or Vinyl lined Pulp Supplied with caps attached in convenience packs.