Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phoenix S&T Inc BTTRFLY PRTFLIO HTER W EASYSPR
SDP

Catalog No. NC3238192
Encompass

  • Insulated heating surfaces.
  • Compatibility with Easy Spray source controller.
  • Temperature stability within 0.2 °C (when controlled by compatible controller).
  • Maximum temperature of 100 °C.
  • Length of 20 cm.
  • Applications in nanoLC-MS, microLC-MS, and nano/microspray.
  • Required accessory: Phoenix S&T Column Heater Controller.
  • Compatible with columns greater than 50 cm long (when coiled).
  • Easy placement of fittings and columns.
  • Contents secured by heat-resistant tape.
  • Placement adjacent to mass spectrometry source.
  • Enhances reproducibility, sensitivity, reduces run pressures, and carry-over.
  • Effective for hydrophobic peptides.

Catalog No. NC3238192 Supplier Phoenix S&T Inc Supplier No. PSTESBPHI
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Insulated heating surfaces.
  • Compatibility with Easy Spray source controller.
  • Temperature stability within 0.2 °C (when controlled by compatible controller).
  • Maximum temperature of 100 °C.
  • Length of 20 cm.
  • Applications in nanoLC-MS, microLC-MS, and nano/microspray.
  • Required accessory: Phoenix S&T Column Heater Controller.
  • Compatible with columns greater than 50 cm long (when coiled).
  • Easy placement of fittings and columns.
  • Contents secured by heat-resistant tape.
  • Placement adjacent to mass spectrometry source.
  • Enhances reproducibility, sensitivity, reduces run pressures, and carry-over.
  • Effective for hydrophobic peptides.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.