Learn More
Vitascientific BTTY OPRTD MINI VORTEX 5600RPM
SDP
Catalog No. NC2046879
Encompass_Preferred
Battery Operated Mini Vortex, 5600 rpm, Orbital diameter: 6mm, Fully charged battery can operate for two hours, each
Catalog No. NC2046879 Supplier Vitascientific Supplier No. FESEC0019
Battery Operated Mini Vortex, 5600 rpm, Orbital diameter: 6mm, Fully charged battery can operate for two hours, each