Learn More
Medchemexpress LLC BTX-A51 (casein kinase inhibitor A51) | 2079068-74-7 | MFCD34469352 | 98.6% | 360.88 | C18H25ClN6 | 100MG
SDP
Catalog No. 5000237382
Encompass_Preferred
BTX-A51 is a potent, orally active inhibitor of casein kinase 1α (CK1α) used in preclinical research on leukemia and CK1α-related signaling pathways. It induces apoptosis in leukemia cell models and is supplied as a research-grade compound with characterized purity, formula, and molecular weight.
  • Potent CK1α inhibition demonstrated in cellular assays.
  • Induces apoptosis in leukemia cell models.
  • Suitable for in vitro and in vivo preclinical studies.
  • Characterized by confirmed molecular formula and molecular weight.
  • Available in multiple milligram to gram quantities for research use.
  • Provided with datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-37382 Supplier Medchemexpress LLC Supplier No. HY123954100MG
BTX-A51 is a potent, orally active inhibitor of casein kinase 1α (CK1α) used in preclinical research on leukemia and CK1α-related signaling pathways. It induces apoptosis in leukemia cell models and is supplied as a research-grade compound with characterized purity, formula, and molecular weight.
  • Potent CK1α inhibition demonstrated in cellular assays.
  • Induces apoptosis in leukemia cell models.
  • Suitable for in vitro and in vivo preclinical studies.
  • Characterized by confirmed molecular formula and molecular weight.
  • Available in multiple milligram to gram quantities for research use.
  • Provided with datasheet, certificate of analysis, and safety data sheet.