Learn More
BTX™ ECM™ 630/Enhancer™ 3000 Electroporation/Monitoring Systems
Catalog No. BTX450071 Shop All BTX Harvard Products
Change view
Click to view available options
ECM™ 630/Enhancer™ 3000 Electroporation/Monitoring Systems; Includes: ECM 630 Generator, Enhancer Interface Probe, Enhancer Interface Box, Digital Oscilloscope, Communications Module Safety Stand, Cuvettes, Cables
Catalog No.
BTX450071 ECM™ 630/Enhancer™ 3000 Electroporation/Monitoring Systems; Includes: ECM 630 Generator, Enhancer Interface Probe, Enhancer Interface Box, Digital Oscilloscope, Communications Module Safety Stand, Cuvettes, Cables
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. BTX450071 Supplier BTX™ Supplier No. 450071
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Includes
  • ECM Generator
  • Enhancer Interface Probe
  • Enhancer Interface Box
  • Digital Oscilloscope
  • Communications Module
  • Safety Stand
  • 30 cuvets (10 each of 1mm, 2mm, and 4mm gap sizes)
  • Cables
ECM™ 630/Enhancer™ 3000 Electroporation/Monitoring Systems combines advanced monitoring capabilities with traditional electroporation generators.
Enhancer 3000 System confirms and track key electroporation parameters, including waveform, peak voltage, field strength, pulse length, pulse duration, and frequency, in all electroporation generators with external pulse capabilities.
  • Display actual electroporation parameters as waveform and verify conditions for optimization and troubleshooting
  • Capture electroporation settings and analyze pulse amplitude, field strength, pulse length, AC pulse duration and frequency
  • Communications module enables user to print or store data for routine quality control, methods validation, and SOP compliance
  • Images can be tracked and downloaded to your computer for analysis
  • Includes: ECM Generator, Enhancer Interface Probe, Enhancer Interface Box, Digital Oscilloscope, Communications Module Safety Stand, 30 Cuvettes (10 Ea. of 1 mm, 2 mm, and 4 mm gap sizes), Cables
Choice of Generators
  • ECM 630, an exponential decay generator recommended for high throughput electroporation of bacterial and yeast samples
  • ECM 830, a square wave generator, for biomolecular screening, transfection, and gene silencing with no cytotoxicity
Specifications
For Use With (Application) Optimize and Troubleshoot Electroporation Settings, Capture and Print Result for Documentation Purpose
Includes ECM 630 Generator, Enhancer Interface Probe, Enhancer Interface Box, Digital Oscilloscope, Communications Module Safety Stand, Cuvettes, Cables