Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ Tweezertrode™ Electrode

Catalog No. p-143895 Shop All BTX Harvard Products
Change view
Click to view available options
Diameter (Metric):
1 mm
3 mm
5 mm
7 mm
10 mm
Material:
Platinum
Stainless Steel
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Material
BTX450486 1 mm Platinum
BTX450487 3 mm Platinum
BTX450489 5 mm Platinum
BTX450488 7 mm Platinum
BTX520 7 mm Stainless Steel
BTX522 10 mm Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. BTX450486 Supplier BTX™ Supplier No. 450486
Only null left
Add to Cart
Add to Cart

Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned

Order Info

Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.

Specifications

For Use With (Application) Vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
For Use With (Equipment) BTX 830, ECM 2001+, Gemini X2 Generators
Material Platinum
Product Line BTX Electroporation
Product Type Tweezer Style Electrode
Voltage 200 V
Description Platinum Tweezertrode, 1 mm Diameter
Diameter (English) 0.039 in.
Diameter (Metric) 1 mm
Electrode Shape Circular
Length (English) Electrode 0.47 in.
Length (Metric) Electrode 11.9 mm
Operating Temperature (English) 41°F to 104°F
Pulse Number Range 1 sec. to 99 msec.
Pulse Length Range 1 μsec. to 200 msec.
Operating Temperature (Metric) 5°C to 40°C
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (Metric) Gap Adjustable: < 1 mm to > 2 cm
Tip Type Tweezer Style
Tweezer Size 11.5 cm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.