Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ Tweezertrode™ Electrode Kit

Catalog No. p-4901617 Shop All BTX Harvard Products
Change view
Click to view available options
Diameter (Metric):
7 mm
10 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
BTX450165 7 mm
BTX450166 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX450165 Supplier BTX™ Supplier No. 450165
Only null left
Add to Cart
Add to Cart

Tweezertrode™ Electrode Kit features reusable, tweezer-style, in vivo electrodes suitable for drug or gene delivery in animals.

  • Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel or platinum at the tip
  • Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
  • Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
  • Can be easily cleaned
  • Kits are supplied with BTX524 coaxial cable for connection to electroporation generators

Specifications

For Use With (Application) In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery.
For Use With (Equipment) ECM 830, Gemini X2, ECM 2001+, Legacy ECM 2001 Generator
Material Stainless Steel
Product Line BTX Electroporation
Product Type Electrode Kit
Voltage 1000 V
Description Stainless Tweezertrode Kit, 7 mm
Diameter (English) 0.27 in.
Diameter (Metric) 7 mm
Electrode Shape Circular
Includes Cable
Length (English) 0.27 in.
Length (English) Electrode 0.47 in.
Length (Metric) 7 mm
Length (Metric) Electrode 12 mm
Pulse Length Range 10 μsec. to 99 msec. in PBS
Quantity 1 Ea.
Relative Humidity 20 to 80%
Size (English) Gap <0.04 in. to >0.79 in.
Size (Metric) Gap <1 mm to >2 cm
Tweezer Size 11.5 cm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.