Learn More
BTX™ Genetrodes™ Holder with Shaft
Catalog No. BTX515 Shop All BTX Harvard Products
Change view
Click to view available options
Connection Type:
Banana Cables
Catalog No. Connection Type
BTX515 Banana Cables
1 options
Catalog No. BTX515 Supplier BTX™ Supplier No. 450203
Add to Cart
Add to Cart
Genetrodes™ Holder with Shaft for use with Genetrodes Electrodes.
  • Correctly positions all electrodes at a predetermined gap (10 grooves, 1 mm apart)
Specifications
For Use With (Application) Correctly Positions Electroporation Electrodes at a Predetermined Gap
For Use With (Equipment) BTX Genetrode or Genepaddle Electrode
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Genetrode/Genepaddle Holder
Includes Handle. Requires Electrodes
Quantity 1 Ea.
Size (English) Gap 0.03 to 0.39 in.
Size (Metric) Gap 1 to 10 mm
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Square Wave
Required Accessories Electrodes
Show More Show Less
Intended for Research use only. Not for use in diagnostic or clinical procedures.