Learn More
BTX™ HT96 Enhancer™ 3000 Electroporation/Monitoring System
Catalog No. BTX450472 Shop All BTX Harvard Products
Change view
Click to view available options
For Use With (Equipment):
ECM 630 Generator
ECM 830 Generator
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
BTX450472 ECM 830 Generator
BTX450471 ECM 630 Generator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTX450472 Supplier BTX™ Supplier No. 450472
Includes
  • ECM Generator
  • HT-200 Plate Handler
  • Two 96-well HT plates
  • Enhancer 3000 Monitoring System with communications module
HT96 Enhancer™ 3000 Electroporation/Monitoring Systems combine rapid multi-well electroporation with customizable data monitoring and analysis for fast, accurate results.
The HT-96 Multi-Well System uses 96-well plates for faster turnaround than traditional cuvets. Run multiple experiments simultaneously, or load each plate with the same sample for high throughput processing. The Enhancer 3000 System enables the user to observe waveform, confirm data, track results, and document key parameters for protocol optimization, troubleshooting, and routine quality control.
  • Processes multiple samples in seconds for optimization of electroporation variables
  • Complete user control over electroporation settings allows for a wide range of optimization flexibility
  • Results are confirmed automatically
  • Monitoring and documentation of waveforms and electroporation parameters lets you quickly determine potential problems
  • Communications module offers standard printer and computer interface
Choice of Generators
  • ECM 630, an exponential decay generator recommended for high throughput electroporation of bacterial and yeast samples
  • ECM 830, a square wave generator, for biomolecular screening, transfection, and gene silencing with no cytotoxicity
Specifications
For Use With (Application) Transformation of Bacteria and Yeast, Transfection of Mammalian Cells
For Use With (Equipment) ECM 830 Generator
Includes ECM Generator, HT-200 Plates Handler, 2 x 96-well HT Plates, Enhancer 3000 Monitoring System with Communications Module