Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer BTX45-0125 2MM CUVETTES,50/PK

Catalog No. NC1900706 Shop All Cole-Parmer Products
Encompass_Preferred

BTX 45-0125 Electroporation Cuvettes, Sterile, 2 mm Gap; 50/Pk

Catalog No. NC1900706 Supplier Cole-Parmer Supplier No. 0289432
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BTX 45-0125 Electroporation Cuvettes, Sterile, 2 mm Gap; 50/Pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.