Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ BTXPress™ Electroporation Buffer Kit

Catalog No. BTX450803 Shop All BTX Harvard Products
Change view
Click to view available options
Quantity:
2 Bags (10 Cuvettes/Bag)
2 Bags (50 Cuvettes/Bag)
4 Bags (10 Cuvettes/Bag), 10 mL Buffer
5 Bags (10 Cuvettes/Bag)
No. of Reactions:
20
40
50
100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity No. of Reactions
BTX450803 5 Bags (10 Cuvettes/Bag) 50
BTX450806 2 Bags (50 Cuvettes/Bag) 100
BTX450804 2 Bags (10 Cuvettes/Bag) 20
BTX450807 4 Bags (10 Cuvettes/Bag), 10 mL Buffer 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BTX450803 Supplier BTX™ Supplier No. 450803
Add to Cart
Add to Cart

BTXPress™ Electroporation Buffer Kit offers a single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered hard to transfect by chemical and other non-viral methods.

BTXPress™ Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you. As a universal solution, the BTXPress™ Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXPress™ Electroporation Solution is offered as a kit including the BTX™ cuvettes Plus with transfer pipettes or as a buffer alone.

  • Efficient electroporation transfection
  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types

Specifications

Content And Storage 2°C to 8°C
Color Clear
Description BTXpress Solution Kit: 50 Reactions in 2 mm gap Cuvettes
Cell Type Mammalian
Sample Type Cells
Promoter None (Promoterless)
Transfection Technique Electroporation
For Use With (Equipment) BTX™ and other Electroporation systems
For Use With (Application) Transfection via Electroporation
Includes 5 mL Solution, 50 x 2 mm Gap Cuvettes and Transfer Pipette
Packaging Type Individually wrapped 2 mm Cuvettes
No. of Reactions 50
Product Line BTX Electroporation
Product Type Electroporation Buffer
Quantity 5 Bags (10 Cuvettes/Bag)
Reagent Type Universal Electroporation Buffer
Sterility Sterile
Volume (Metric) 5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.