Learn More
Medchemexpress LLC HY-13579A 5mg Medchemexpress, BTZ043 Racemate CAS:957217-65-1 Purity:>98%
SDP
Catalog No. 501932665
Encompass_Preferred
Medchemexpress, HY-13579A 5mg BTZ043 Racemate CAS:957217-65-1 BTZ043 Racemate is the racemate of BTZ043, BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase ( DprE1 ), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2665 Supplier Medchemexpress LLC Supplier No. HY13579A5MG
Medchemexpress, HY-13579A 5mg BTZ043 Racemate CAS:957217-65-1 BTZ043 Racemate is the racemate of BTZ043, BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase ( DprE1 ), and the antimicrobial activity of BTZ043 is more potent than BTZ043 Racemate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.