Learn More
Cayman Chemical BU 224hydrochlorIde 10mg
SDP
Catalog No. 502945182
Encompass_Preferred
An imidazoline receptor agonist; selectively binds to imidazoline I1 and -I2 receptors over a2-ARs (IC50s = 0.034, 0.026, and 15 µM, respectively); reduces complete Freund’s adjuvant-induced thermal and mechanical hyperalgesia in rats at 10 or 17.8 mg/kg; decreases the latency to seizure onset and increases the percentage of animals with grade V seizures to a greater extent in female than male mice; reverses cognitive deficits in the 5XFAD transgenic mouse model of Alzheimer’s disease at 5 mg/kg
Catalog No. 50-294-5182 Supplier Cayman Chemical Supplier No. 4245110 MG
An imidazoline receptor agonist; selectively binds to imidazoline I1 and -I2 receptors over a2-ARs (IC50s = 0.034, 0.026, and 15 µM, respectively); reduces complete Freund’s adjuvant-induced thermal and mechanical hyperalgesia in rats at 10 or 17.8 mg/kg; decreases the latency to seizure onset and increases the percentage of animals with grade V seizures to a greater extent in female than male mice; reverses cognitive deficits in the 5XFAD transgenic mouse model of Alzheimer’s disease at 5 mg/kg