Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BU224 hydrochloride | 205437-64-5 | MFCD00467639 | 97.0% | 233.70 g/mol | C12H12ClN3 | 1 ML
SDP

Catalog No. 5000211233
Encompass_Preferred

BU224 hydrochloride is the hydrochloride salt of an imidazoline I2 receptor ligand supplied as a ready-to-use 10 mM solution (1 mL) in DMSO for laboratory research. It is used in neuroscience studies of I2 receptor pharmacology and neuroprotection, including preclinical models of cognitive impairment.

  • Provided as a 10 mM solution, 1 mL.
  • Solvent: DMSO.
  • Purity: 96.96%.
  • Molecular weight: 233.70 g/mol; formula C12H12ClN3.
  • Form: brown to gray solid (solid characterization reported).
  • Storage: solid at 4°C protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month) protected from light.

Catalog No. 50-002-11233 Supplier Medchemexpress LLC Supplier No. HY10117010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BU224 hydrochloride is the hydrochloride salt of an imidazoline I2 receptor ligand supplied as a ready-to-use 10 mM solution (1 mL) in DMSO for laboratory research. It is used in neuroscience studies of I2 receptor pharmacology and neuroprotection, including preclinical models of cognitive impairment.

  • Provided as a 10 mM solution, 1 mL.
  • Solvent: DMSO.
  • Purity: 96.96%.
  • Molecular weight: 233.70 g/mol; formula C12H12ClN3.
  • Form: brown to gray solid (solid characterization reported).
  • Storage: solid at 4°C protected from light; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month) protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.