Learn More
Sino Biological Recombinant Human BRAF/B-Raf Protein (GST Tag), Active
Catalog No. 502575828
Encompass_Preferred
Recombinant human BRAF (381-end or exon 9-18) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5828 Supplier Sino Biological Supplier No. B0811BG500
Recombinant human BRAF (381-end or exon 9-18) protein was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.