Learn More
Aviva Systems Biology Corp BUB3 Antibody - N-terminal region (ARP58599_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58599_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BUB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BUB3. Alternate names: BUB3L, hBUB3ApplicationWB, IF, IHC. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62367