Learn More
Tinius Olsen Testing Machine Bubble Level Assembly for MFI
SDP
Catalog No. NC3800217
Encompass
Use this tool to ensure the furnace cylinder is level as it sits on your lab benchtop. Simply place the orifice and piston rod in the cylinder with the level sitting on top of the exposed end of the piston rod.
Catalog No. NC3800217 Supplier Tinius Olsen Testing Machine Supplier No. 02001271
Use this tool to ensure the furnace cylinder is level as it sits on your lab benchtop. Simply place the orifice and piston rod in the cylinder with the level sitting on top of the exposed end of the piston rod.