Learn More
Hach Company Bubble Trap Head Regulator
Supplier:  Hach Company 4668000
Encompass
When installed the bubble trap and head regulator for the 1720 family of turbidimeters works with the internal instrument bubble trap to remove bubbles and also helps to regulate flow to an optimal speed in order to get the best flow going into the turbidimeter
Catalog No. 50-002-07067