Learn More
Ace Glass, Inc. BUBBLER BUBBLER
SDP
Catalog No. 50975469
Encompass_Preferred
Bubbler-Manometer for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-469 Supplier Ace Glass, Inc. Supplier No. 781839
Bubbler-Manometer for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.