Learn More
Chemglass Life Sciences Bubbler, Double, Airfree, Schlenk
SDP
Catalog No. AF0513D21
Encompass_Preferred
Bubbler, Double, Airfree, Schlenk. Similar in design to AF-0513 but with two oil reservoirs. Bubbler is approx 125mm W x 135mm H. Hose connections are 9mm OD at the middle serration.
Catalog No. AF0513D21 Supplier Chemglass Life Sciences Supplier No. AF0513D21
Bubbler, Double, Airfree, Schlenk. Similar in design to AF-0513 but with two oil reservoirs. Bubbler is approx 125mm W x 135mm H. Hose connections are 9mm OD at the middle serration.