Learn More
Ace Glass, Inc. BUBBLER AND TRAP BUBBLER AND TRAP
SDP
Catalog No. 50975471
Encompass_Preferred
Bubbler and Trap with #7 Ace-Thred, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.
Catalog No. 50-975-471 Supplier Ace Glass, Inc. Supplier No. 781846
Bubbler and Trap with #7 Ace-Thred, for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.