Learn More
Chemglass Life Sciences Bubbler, Cajon, Connection, Airfree, Schlenk
SDP
Catalog No. AF008501
Encompass_Preferred
Bubbler, Cajon, Connection, Airfree, Schlenk. Bubbler has an integral valve to control the flow of inert gas into the system. Bubbler requires a minimal amount of oil due to the inner tube being within 5mm of the bottom. Both sidearms are 9mm OD for use with 3/8" Cajon fittings. Overall height is approx. 220mm.
Catalog No. AF008501 Supplier Chemglass Life Sciences Supplier No. AF008501
Bubbler, Cajon, Connection, Airfree, Schlenk. Bubbler has an integral valve to control the flow of inert gas into the system. Bubbler requires a minimal amount of oil due to the inner tube being within 5mm of the bottom. Both sidearms are 9mm OD for use with 3/8" Cajon fittings. Overall height is approx. 220mm.