Learn More
Chemglass Life Sciences Bubbler, Horizontal, 14/20 Inner Joint, Airfree, Schlenk
SDP
Catalog No. 50872947
Encompass_Preferred
Bubbler, Horizontal, 14/20 Inner Joint, Airfree, Schlenk. Horizontal design of bubbler allows for in line connection and purging of apparatus. Hose connection has a 9mm OD middle serration. Complete with standard taper inner joint.
Catalog No. 50-872-947 Supplier Chemglass Life Sciences Supplier No. AF051401
Bubbler, Horizontal, 14/20 Inner Joint, Airfree, Schlenk. Horizontal design of bubbler allows for in line connection and purging of apparatus. Hose connection has a 9mm OD middle serration. Complete with standard taper inner joint.