Learn More
Ace Glass, Inc. Bubbler, No-Air, metering valve, 9mm OD tube-end sidearms
SDP
Catalog No. NC0372040
Encompass_Preferred
Bubbler with metering valve for precise control of inert gas introduction and features 9mm OD tube-end side arms for use with 3/8in ID vacuum tubing.
Catalog No. NC0372040 Supplier Ace Glass, Inc. Supplier No. 741301
Bubbler with metering valve for precise control of inert gas introduction and features 9mm OD tube-end side arms for use with 3/8in ID vacuum tubing.