Learn More
Wilmad Labglass BUBBLER MINERAL OIL 8MM
Catalog No. 50806424
Encompass_Preferred
Bubbler, Mineral Oil. Used to observe gaseous additions to reactions. May be used to vent a reaction mixture to atmosphere. Tubing connections are 8mm OD.
Catalog No. 50-806-424 Supplier Wilmad Labglass Supplier No. IG145100
Bubbler, Mineral Oil. Used to observe gaseous additions to reactions. May be used to vent a reaction mixture to atmosphere. Tubing connections are 8mm OD.