Learn More
Chemglass Life Sciences Bubbler, Modified, 14/20 Inner Joint, 130mm OAH, Airfree, Schlenk
SDP
Catalog No. 501530965
Encompass_Preferred
Bubbler, Modified, 14/20 Inner Joint, 130mm OAH, Airfree, Schlenk. Used to monitor gas flow to reactions while providing a vent to the atmosphere. Reservoir head prevents oil from being sucked back into the system. Standard taper inner joint is for use in single or multi-neck flasks. Hose connection has a 9mm OD middle serration.
Catalog No. 50-153-0965 Supplier Chemglass Life Sciences Supplier No. AF051601
Bubbler, Modified, 14/20 Inner Joint, 130mm OAH, Airfree, Schlenk. Used to monitor gas flow to reactions while providing a vent to the atmosphere. Reservoir head prevents oil from being sucked back into the system. Standard taper inner joint is for use in single or multi-neck flasks. Hose connection has a 9mm OD middle serration.