Learn More
Chemglass Life Sciences Bubbler, Modified, 24/40 Inner Joint, 150mm OAH, Airfree, Schlenk
SDP
Catalog No. AF051602
Encompass_Preferred
Bubbler, Modified, 24/40 Inner Joint, 150mm OAH, Airfree, Schlenk. Used to monitor gas flow to reactions while providing a vent to the atmosphere. Reservoir head prevents oil from being sucked back into the system. Standard taper inner joint is for use in single or multi-neck flasks. Hose connection has a 9mm OD middle serration. The 24/40 jointed bubbler is available in two sizes; the larger for use in vessels having a capacity greater than 100mL.
Catalog No. AF051602 Supplier Chemglass Life Sciences Supplier No. AF051602
Bubbler, Modified, 24/40 Inner Joint, 150mm OAH, Airfree, Schlenk. Used to monitor gas flow to reactions while providing a vent to the atmosphere. Reservoir head prevents oil from being sucked back into the system. Standard taper inner joint is for use in single or multi-neck flasks. Hose connection has a 9mm OD middle serration. The 24/40 jointed bubbler is available in two sizes; the larger for use in vessels having a capacity greater than 100mL.