Learn More
Chemglass Life Sciences Bubbler, Pressure Release, 130mm Height x 100mm Width x 100mm Depth
SDP
Catalog No. CG453601
Encompass_Preferred
Bubbler, Pressure Release, 130mm Height x 100mm Width x 100mm Depth. Used for the purging of AIRFREE or vacuum apparatus with an inert gas. The pressure release valve is connected on one side to the low pressure gas source and the other side to a manifold or reaction vessel. The check valve prevents oil and air from being pulled into the apparatus. Hose connections are 10mm at the largest serration.
Catalog No. CG453601 Supplier Chemglass Life Sciences Supplier No. CG453601
Bubbler, Pressure Release, 130mm Height x 100mm Width x 100mm Depth. Used for the purging of AIRFREE or vacuum apparatus with an inert gas. The pressure release valve is connected on one side to the low pressure gas source and the other side to a manifold or reaction vessel. The check valve prevents oil and air from being pulled into the apparatus. Hose connections are 10mm at the largest serration.