Learn More
Boca Scientific Inc BuccalFix Plus DNA Isolation Kit, 50 reactions
Supplier:  Boca Scientific Inc BFP50
Encompass
The BuccalFix Plus DNA Isolation Kit is designed to easily and quickly purify DNA already stored in BuccalFix Stabilization buffer. The kit comes with all the reagents required to perform the necessary isolation steps, including Proteinase K to produce DNA with high yields and purity. The protocol is extremely flexible and easily adjusted if there are varying volumes to be purified. The average processing time for 0.5 ml is under an hour, with no solvents, alcohol or other buffers required.
Catalog No. NC1800041