Learn More
Boca Scientific Inc BuccalFix DNA/RNA Stabilization Buffer, 25ml, Sufficient for Stabilizing 50 swabs
Catalog No. NC1800039
Encompass
BuccalFix Stabilization Buffer is designed to completely stabilize the DNA and RNA on buccal swabs by inhibiting all enzymatic and microbial activity that occurs following buccal sampling. DNA can be stored in BuccalFix at ambient temperature for over 2 years, while RNA can be stabilized for 4 days. While stabilized, the structure and integrity of the DNA and RNA is fully maintained for further downstream processing applications. Stabilized DNA in BuccalFix can be processed for isolation by the addition of Proteinase K and subsequent isolation steps.
Catalog No. NC1800039 Supplier Boca Scientific Inc Supplier No. BFX25
BuccalFix Stabilization Buffer is designed to completely stabilize the DNA and RNA on buccal swabs by inhibiting all enzymatic and microbial activity that occurs following buccal sampling. DNA can be stored in BuccalFix at ambient temperature for over 2 years, while RNA can be stabilized for 4 days. While stabilized, the structure and integrity of the DNA and RNA is fully maintained for further downstream processing applications. Stabilized DNA in BuccalFix can be processed for isolation by the addition of Proteinase K and subsequent isolation steps.