Learn More
Cayman Chemical BuccalIn AtrIfluoroacetAT 10mg
SDP
Catalog No. 502919283
Encompass_Preferred
A neuropeptide; reduces current-induced muscle contraction size but not muscle relaxation rate in isolated and washed A. californica ARC muscle fibers at 10 nM; decreases the amplitude of inhibitory post-synaptic currents (IPSCs) in isolated and perfused voltage-clamped A. californica buccal ganglia at 10 µM
Catalog No. 50-291-9283 Supplier Cayman Chemical Supplier No. 3972810 MG
A neuropeptide; reduces current-induced muscle contraction size but not muscle relaxation rate in isolated and washed A. californica ARC muscle fibers at 10 nM; decreases the amplitude of inhibitory post-synaptic currents (IPSCs) in isolated and perfused voltage-clamped A. californica buccal ganglia at 10 µM