Learn More
BUCHI Rotavapor Bump Trap SJ 24/40
SDP
Catalog No. 10987130 Shop All BUCHI Products
Change view
Click to view available options
Taper:
24/40
Catalog No. Taper
10-987-130 24/40
1 options
Catalog No. 10-987-130 Supplier BUCHI Supplier No. 11056919
Add to Cart
Add to Cart
Bump trap adapter.
  • BUCHI glassware
  • Glass adapters for excessively foaming samples
  • Prevents foam from entering the vapor duct and condenser
  • Glass, SJ24/40, 175mm
Specifications
Material Borosilicate Glass
Type Bump Trap
For Use With (Application) Modifies vapor pathway in event of bumping or foaming
For Use With (Equipment) Rotary evaporator
Taper 24/40