Learn More
BUCHI Rotavapor Fixing Spring
SDP
Catalog No. 05422522 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
Rotary evaporator
Catalog No. For Use With (Equipment)
05-422-522 Rotary evaporator
1 options
Catalog No. 05-422-522 Supplier BUCHI Supplier No. 000633
Add to Cart
Add to Cart
The fixing spring is an internal component that holds the vapor duct securely in the drive unit of BUCHI Rotavapors.
  • Designed for Rotavapor
  • Secures vapor duct
Specifications
Type Fixing Spring with Metal Clip
For Use With (Application) Holds vapor duct in place
For Use With (Equipment) Rotary evaporator
Holds Vapor Duct