Learn More
BUCHI Beaker-Style Drying Flasks for Rotary Evaporators
SDP
Catalog No. 05404321 Shop All BUCHI Products
Change view
Click to view available options
Taper:
24/40
Catalog No. Taper
05-404-321 24/40
1 options
Catalog No. 05-404-321 Supplier BUCHI Supplier No. 034230
Specifications
Material Standard Glass
Type Drying Flask
For Use With (Equipment) Rotary evaporator
Material (Coating) Plastic and Glass
Shape Pear
Taper 24/40
Temperature (Metric) Operating 60°C
Volume (Metric) 500 mL